1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane ≥94.0%
Supplier: TCI America
B3319-25G
B3319-5G
TCB3319-25GEA
431.34
CAD
TCB3319-25G
TCB3319-5G
1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane ≥94.0%
1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane
CAS Number: 39482-87-6
MDL Number: MFCD00153070
Molecular Formula: C10H21F6NSi2
Molecular Weight: 325.45
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 78
Specific Gravity (20/20): 1.10
Formula:
C₁₀H₂₁F₆NSi₂ Boiling Pt: 119 °C Density: 1.10 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD00153070 CAS Number: 39482-87-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...