(R)-1-Benzyl-3-pyrrolidinecarbonitrile ≥98.0%

Supplier: TCI America

B3299-5G B3299-1G
TCB3299-5GEA 1213.28 CAD
TCB3299-5G TCB3299-1G
(R)-1-Benzyl-3-pyrrolidinecarbonitrile ≥98.0%
(R)-1-Benzyl-3-pyrrolidinecarbonitrile

CAS Number: 157528-56-8
MDL Number: MFCD09951967
Molecular Formula: C12H14N2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific rotation [a]20/D: -30 deg (C=10, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Benzyl-3-cyanopyrrolidine

Formula: C₁₂H₁₄N₂
MDL Number: MFCD09951967
CAS Number: 157528-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7c0d5a2f86925ed24906e4a5392fe39ee03cc0&vid=5e7c0d5a2f86925ed24906e4a5392fe39ee03cc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR