Butenafine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
N-(4-(tert-Butyl)benzyl)-N-methyl-1-(naphthalen-1-yl)methanamine hydrochloride, Butenafine HCl
B3293-5G
B3293-1G
TCB3293-5GEA
565.64
CAD
TCB3293-5G
TCB3293-1G
Butenafine hydrochloride ≥98.0%
Butenafine hydrochloride
CAS Number: 101827-46-7
MDL Number: MFCD00917064
Molecular Formula: C23H27N
Molecular Weight: 353.93
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210
Formula:
C₂₃H₂₇N MW: 353.93 g/mol Melting Pt: 210 °C |
MDL Number:
MFCD00917064 CAS Number: 101827-46-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...