Cysteamine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

A0648-25G A0648-500G
TCA0648-025GEA 107.27 CAD
TCA0648-025G TCA0648-500G
Cysteamine ≥95.0% (by titrimetric analysis)
Cysteamine

CAS Number: 60-23-1
MDL Number: MFCD00008196
Molecular Formula: C2H7NS
Molecular Weight: 77.15
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 95
Storage Temperature: 0-10°C


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Antioxidants, Crosslinkers
Other Product Names / Synonyms: Cysteamine, 2-Mercaptoethylamine

Formula: C₂H₇NS
Melting Pt: 95 °C
MDL Number: MFCD00008196
CAS Number: 60-23-1
EINECS: 205-858-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a55bfc80f3fcb7b12f93fd0642f2cda306a316&vid=87a55bfc80f3fcb7b12f93fd0642f2cda306a316&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR