1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97.0%

Supplier: TCI America

Synonyms: 1,2-Bis(phenylsulphinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulfinyl)ethane palladium(II) acetate, 1,2-Bis(phenylsulphinyl)ethane palladium(II) acetate, White Catalyst, Christina White Catalyst

B3292-1G B3292-200MG
TCB3292-1GEA 286.99 CAD
TCB3292-1G TCB3292-200MG
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97.0%
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate

CAS Number: 858971-43-4
MDL Number: MFCD09842752
Molecular Formula: C18H20O6PdS2
Molecular Weight: 502.89
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction
Other Product Names / Synonyms: White Catalyst

Formula: C₁₆H₁₈O₄PdS₂
MW: 444.86 g/mol
MDL Number: MFCD09842752
CAS Number: 858971-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fc8089227bfb931dcaba3ff544f1c2bc3c30040&vid=2fc8089227bfb931dcaba3ff544f1c2bc3c30040&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR