2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3291-25G B3291-5G
TCB3291-25GEA 154.84 CAD
TCB3291-25G TCB3291-5G
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride ≥98.0% (by HPLC, titration analysis)
2-Butyl-1,3-diazaspiro[4.4]non-1-en-4-one hydrochloride

CAS Number: 151257-01-1
MDL Number: MFCD03428357
Molecular Formula: C11H18N2O
Molecular Weight: 230.74
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₁H₁₈N₂O
MW: 230.74 g/mol
MDL Number: MFCD03428357
CAS Number: 151257-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=210b9dfad0a176e76459f048bf62d449e2620aca&vid=210b9dfad0a176e76459f048bf62d449e2620aca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR