4-Bromotetraphenylsilane ≥96.0%
Supplier: TCI America
B3287-1G
B3287-200MG
TCB3287-1GEA
140.62
CAD
TCB3287-1G
TCB3287-200MG
4-Bromotetraphenylsilane ≥96.0%
4-Bromotetraphenylsilane
CAS Number: 18737-40-1
Molecular Formula: C24H19BrSi
Molecular Weight: 415.41
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 175
Formula:
C₂₄H₁₉BrSi Melting Pt: 175 °C |
MDL Number:
MFCD00093371 CAS Number: 18737-40-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...