N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol ≥93.0%

Supplier: TCI America

Synonyms: 2-(tert-Butoxycarbonylamino)-2-phenylethanol, N-Boc-DL-2-phenylglycinol, Boc-DL-Phenylglycinol, tert-Butyl (2-hydroxy-1-phenylethyl)carbamate

B3271-1G B3271-5G
TCB3271-1GEA 45.82 CAD
TCB3271-1G TCB3271-5G
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol ≥93.0%
N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol

CAS Number: 67341-01-9
MDL Number: MFCD01883039
Molecular Formula: C13H19NO3
Molecular Weight: 237.30
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 142


Other Product Names / Synonyms: N-Boc-DL-2-phenylglycinol, 2-(tert-Butoxycarbonylamino)-2-phenylethanol

Formula: C₁₃H₁₉NO₃
Melting Pt: 142 °C
MDL Number: MFCD01883039
CAS Number: 67341-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c5bb36a643eedc3127547fa1c03b3c36d556667&vid=2c5bb36a643eedc3127547fa1c03b3c36d556667&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR