trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3253-25G
B3253-5G
TCB3253-25GEA
785.1
CAD
TCB3253-25G
TCB3253-5G
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid
CAS Number: 27687-14-5
Molecular Formula: C13H23NO4
Molecular Weight: 257.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 135
Other Product Names / Synonyms: trans-4-(Boc-aminomethyl)cyclohexanecarboxylic Acid
Formula:
C₁₃H₂₃NO₄ Melting Pt: 135 °C |
MDL Number:
MFCD00673773 CAS Number: 27687-14-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...