Ammelide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Cyanuramide, Melanuric Acid, Dihydro-6-imino-1,3,5-triazine-2,4(1H,3H)-dione, 6-Amino-1,3,5-triazine-2,4-diol, Cyanuric Acid Monoamide
A0645-1G
A0645-5G
TCA0645-001GEA
275.79
CAD
TCA0645-001G
TCA0645-005G
Ammelide ≥98.0% (by titrimetric analysis)
Ammelide
CAS Number: 645-93-2
MDL Number: MFCD00049046
Molecular Formula: C3H4N4O2
Molecular Weight: 128.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 6-Amino-1,3,5-triazine-2,4-diol, Cyanuramide, Cyanuric Acid Monoamide, Dihydro-6-imino-1,3,5-triazine-2,4(1H,3H)-dione, Melanuric Acid
Formula:
C₃H₄N₄O₂ MW: 128.09 g/mol |
MDL Number:
MFCD00049046 CAS Number: 645-93-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...