trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3250-5G B3250-25G
TCB3250-5GEA 224.36 CAD
TCB3250-5G TCB3250-25G
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

CAS Number: 53292-89-0
MDL Number: MFCD01862293
Molecular Formula: C12H21NO4
Molecular Weight: 243.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 183


Other Product Names / Synonyms: trans-4-(Boc-amino)cyclohexanecarboxylic Acid

Formula: C₁₂H₂₁NO₄
Melting Pt: 183 °C
MDL Number: MFCD01862293
CAS Number: 53292-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8da0baf0f05fbfb338b0835008080ab6d13d0b0&vid=e8da0baf0f05fbfb338b0835008080ab6d13d0b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR