cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: cis-4-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, cis-4-(BOC-amino)cyclohexanecarboxylic acid, cis-4-(Boc-amino)cyclohexanecarboxylic Acid

B3249-5G B3249-25G
TCB3249-5GEA 130.51 CAD
TCB3249-5G TCB3249-25G
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

CAS Number: 53292-90-3
MDL Number: MFCD01862294
Molecular Formula: C12H21NO4
Molecular Weight: 243.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: cis-4-(Boc-amino)cyclohexanecarboxylic Acid

Formula: C₁₂H₂₁NO₄
MW: 243.30 g/mol
Melting Pt: 169 °C
MDL Number: MFCD01862294
CAS Number: 53292-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32a7a7ccb369cd83e621223e677c127ca45f7bcc&vid=32a7a7ccb369cd83e621223e677c127ca45f7bcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR