4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3248-5G
B3248-25G
TCB3248-5GEA
203.82
CAD
TCB3248-5G
TCB3248-25G
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture)
CAS Number: 130309-46-5
Molecular Formula: C12H21NO4
Molecular Weight: 243.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: 4-(Boc-amino)cyclohexanecarboxylic Acid
Formula:
C₁₂H₂₁NO₄ |
MDL Number:
MFCD03453262 CAS Number: 130309-46-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...