N-Benzyl-N,N'-dimethylethylenediamine ≥98.0%

Supplier: TCI America

B3246-25G B3246-5G
TCB3246-25GEA 794.74 CAD
TCB3246-25G TCB3246-5G
N-Benzyl-N,N'-dimethylethylenediamine ≥98.0%
N-Benzyl-N,N'-dimethylethylenediamine

CAS Number: 102-11-4
MDL Number: MFCD00014856
Molecular Formula: C11H18N2
Molecular Weight: 178.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 74
Specific Gravity (20/20): 0.94

Formula: C₁₁H₁₈N₂
Boiling Pt: 74 °C
Density: 0.94 (20 °C)
MDL Number: MFCD00014856
CAS Number: 102-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=292b5317f8f47cbf8bbde165a4f9c14402f7a9a5&vid=292b5317f8f47cbf8bbde165a4f9c14402f7a9a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR