O-Benzoyl-N-methylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B3239-1G B3239-5G
TCB3239-1GEA 85.32 CAD
TCB3239-1G TCB3239-5G
O-Benzoyl-N-methylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
O-Benzoyl-N-methylhydroxylamine hydrochloride

CAS Number: 27130-46-7
Molecular Formula: C8H9NO2
Molecular Weight: 187.62
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 136

Formula: C₈H₉NO₂
Melting Pt: 136 °C
CAS Number: 27130-46-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc163ab52c34c3da5246a09bc27ab5c06f7e0905&vid=dc163ab52c34c3da5246a09bc27ab5c06f7e0905&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR