O-Benzoyl-N-tert-butylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B3238-1G B3238-5G
TCB3238-1GEA 114.08 CAD
TCB3238-1G TCB3238-5G
O-Benzoyl-N-tert-butylhydroxylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
O-Benzoyl-N-tert-butylhydroxylamine hydrochloride

CAS Number: 66809-86-7
Molecular Formula: C11H15NO2
Molecular Weight: 229.70
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 187

Formula: C₁₁H₁₅NO₂
Melting Pt: 187 °C
CAS Number: 66809-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c01b6badfec812dc2c38bc972c743034b3c2125&vid=4c01b6badfec812dc2c38bc972c743034b3c2125&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR