[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]triphenylphosphine nickel(II) dichloride ≥98.0%

Supplier: TCI America

B3235-1G B3235-200MG
TCB3235-1GEA 436.16 CAD
TCB3235-1G TCB3235-200MG
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]triphenylphosphine nickel(II) dichloride ≥98.0%
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]triphenylphosphine nickel(II) dichloride

CAS Number: 903592-98-3
Molecular Formula: C45H51Cl2N2NiP
Molecular Weight: 780.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₄₅H₅₁Cl₂N₂NiP
MDL Number: MFCD12545958
CAS Number: 903592-98-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18faab264f0a2234c402a1182a02d135a3bed2c7&vid=18faab264f0a2234c402a1182a02d135a3bed2c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR