Benzyl-2,2,2-Trifluoro-N-phenylacetimidate ≥98.0%
Supplier: TCI America
Synonyms:
2,2,2-Trifluoro-N-phenylacetimidic Acid Benzyl Ester
B3234-1G
B3234-5G
TCB3234-1GEA
153.26
CAD
TCB3234-1G
TCB3234-5G
Benzyl-2,2,2-Trifluoro-N-phenylacetimidate ≥98.0%
Benzyl-2,2,2-Trifluoro-N-phenylacetimidate
CAS Number: 952057-61-3
Molecular Formula: C15H12F3NO
Molecular Weight: 279.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.21
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 2,2,2-Trifluoro-N-phenylacetimidic Acid Benzyl Ester
Formula:
C₁₅H₁₂F₃NO MW: 279.26 g/mol Density: 1.21 (20 °C) |
CAS Number:
952057-61-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...