4-Bromo-3-hydroxy-2-naphthoic acid ≥97.0%

Supplier: TCI America

B3229-5G B3229-25G
TCB3229-5GEA 186.76 CAD
TCB3229-5G TCB3229-25G
4-Bromo-3-hydroxy-2-naphthoic acid ≥97.0%
4-Bromo-3-hydroxy-2-naphthoic acid

CAS Number: 2208-15-3
Molecular Formula: C11H7BrO3
Molecular Weight: 267.08
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 231

Formula: C₁₁H₇BrO₃
Melting Pt: 231 °C
MDL Number: MFCD00094166
CAS Number: 2208-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b707aff986357d17590716358216b10b97a67b6f&vid=b707aff986357d17590716358216b10b97a67b6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR