2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Flecainide Impurity D

B3227-25G B3227-5G
TCB3227-25GEA 80.58 CAD
TCB3227-25G TCB3227-5G
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid ≥98.0% (by GC, titration analysis)
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid

CAS Number: 35480-52-5
MDL Number: MFCD00221424
Molecular Formula: C11H8F6O4
Molecular Weight: 318.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123

Formula: C₁₁H₈F₆O₄
MW: 318.17 g/mol
Melting Pt: 123 °C
MDL Number: MFCD00221424
CAS Number: 35480-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf8ea43ef04a02b37aa78522de4a17a1d5172fbe&vid=bf8ea43ef04a02b37aa78522de4a17a1d5172fbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR