3-Bromothieno[3,2-b]thiophene ≥94.0%

Supplier: TCI America

B3226-200MG B3226-1G
TCB3226-200MGEA 402.9 CAD
TCB3226-200MG TCB3226-1G
3-Bromothieno[3,2-b]thiophene ≥94.0%
3-Bromothieno[3,2-b]thiophene

CAS Number: 25121-83-9
Molecular Formula: C6H3BrS2
Molecular Weight: 219.11
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.78
Storage Temperature: 0-10°C

Formula: C₆H₃BrS₂
Density: 1.78 (20 °C)
MDL Number: MFCD10000952
CAS Number: 25121-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39f99f253ed04b6d7fd7e94626737925e7c9ef6a&vid=39f99f253ed04b6d7fd7e94626737925e7c9ef6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR