3-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3221-5G B3221-25G
TCB3221-5GEA 121.66 CAD
TCB3221-5G TCB3221-25G
3-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)
3-Bromo-o-toluic acid

CAS Number: 76006-33-2
MDL Number: MFCD00270097
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 154


Other Product Names / Synonyms: 3-Bromo-o-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 154 °C
MDL Number: MFCD00270097
CAS Number: 76006-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=164be1469eae66e7a26f16385420b08931240ee8&vid=164be1469eae66e7a26f16385420b08931240ee8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR