4'-Bromo-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
B3219-1G
B3219-5G
TCB3219-1GEA
829.5
CAD
TCB3219-1G
TCB3219-5G
4'-Bromo-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)
4'-Bromo-2,2':6',2''-terpyridine
CAS Number: 149817-62-9
MDL Number: MFCD06796965
Molecular Formula: C15H10BrN3
Molecular Weight: 312.17
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 139
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 4'-Bromo-alpha,alpha',alpha''-tripyridyl
Formula:
C₁₅H₁₀BrN₃ Melting Pt: 139 °C |
MDL Number:
MFCD06796965 CAS Number: 149817-62-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...