4'-(4-Bromophenyl)-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4'-(4-Bromophenyl)-alpha,alpha',alpha''-tripyridyl, 4'-(4-Bromophenyl)-2,6':2',2''-terpyridine

B3218-5G B3218-1G
TCB3218-5GEA 560.9 CAD
TCB3218-5G TCB3218-1G
4'-(4-Bromophenyl)-2,2':6',2''-terpyridine ≥97.0% (by HPLC, titration analysis)
4'-(4-Bromophenyl)-2,2':6',2''-terpyridine

CAS Number: 89972-76-9
MDL Number: MFCD06796984
Molecular Formula: C21H14BrN3
Molecular Weight: 388.27
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 162


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 4'-(4-Bromophenyl)-alpha,alpha',alpha''-tripyridyl

Formula: C₂₁H₁₄BrN₃
MW: 388.27 g/mol
Melting Pt: 162 °C
MDL Number: MFCD06796984
CAS Number: 89972-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e3991a699387ff17428c2b653f01f4643ba864&vid=52e3991a699387ff17428c2b653f01f4643ba864&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR