[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers) ≥97.0%
Supplier: TCI America
B3214-5G
B3214-1G
TCB3214-5GEA
376.04
CAD
TCB3214-5G
TCB3214-1G
[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers) ≥97.0%
[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers)
CAS Number: 18401-43-9
MDL Number: MFCD00167591
Molecular Formula: C13H24O3Si
Molecular Weight: 256.42
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 98
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 5-(Triethoxysilyl)-2-norbornene, 5-(Triethoxysilyl)bicyclo[2.2.1]hept-2-ene
Formula:
C₁₃H₂₄O₃Si Boiling Pt: 104 °C Density: 0.99 (20 °C) Flash Pt: 98 °C |
MDL Number:
MFCD00167591 CAS Number: 18401-43-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...