2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥90.0% (by GC, titration analysis)
Supplier: TCI America
B3199-5G
B3199-1G
TCB3199-5GEA
92.59
CAD
TCB3199-5G
TCB3199-1G
2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥90.0% (by GC, titration analysis)
2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS Number: 166330-03-6
Molecular Formula: C7H14BBrO2
Molecular Weight: 220.90
Purity/Analysis Method: >90.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 68
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: (Bromomethyl)boronic Acid Pinacol Ester
Formula:
C₇H₁₄BBrO₂ Boiling Pt: 68 °C Density: 1.26 (20 °C) |
MDL Number:
MFCD09271779 CAS Number: 166330-03-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...