(S,S'')-2,2''-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
B3197-100MG
B3197-500MG
TCB3197-100MGEA
205.4
CAD
TCB3197-100MG
TCB3197-500MG
(S,S'')-2,2''-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
(S,S'')-2,2''-Bis[(S)-4-isopropyloxazolin-2-yl]-1,1''-biferrocene
CAS Number: 190601-12-8
Molecular Formula: C32H36Fe2N2O2
Molecular Weight: 592.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₃₂H₃₆Fe₂N₂O₂ MW: 592.34 g/mol |
CAS Number:
190601-12-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...