(R,R'')-2,2''-Bis(diphenylphosphino)-1,1''-biferrocene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B3196-500MG B3196-100MG
TCB3196-500MGEA 1488.36 CAD
TCB3196-500MG TCB3196-100MG
(R,R'')-2,2''-Bis(diphenylphosphino)-1,1''-biferrocene ≥98.0% (by titrimetric analysis)
(R,R'')-2,2''-Bis(diphenylphosphino)-1,1''-biferrocene

CAS Number: 136274-57-2
Molecular Formula: C44H36Fe2P2
Molecular Weight: 738.41
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 232
Specific rotation [a]20/D: -159 deg (C=0.2, CHCl3)

Formula: C₄₄H₃₆Fe₂P₂
Melting Pt: 232 °C
CAS Number: 136274-57-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ab202d1eece3e930a418c330f00eba38fcdf411&vid=0ab202d1eece3e930a418c330f00eba38fcdf411&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR