4-Bromo-o-phenylenediamine ≥96.0%

Supplier: TCI America

B3181-5G B3181-25G
TCB3181-5GEA 74.26 CAD
TCB3181-5G TCB3181-25G
4-Bromo-o-phenylenediamine ≥96.0%
4-Bromo-o-phenylenediamine

CAS Number: 1575-37-7
MDL Number: MFCD02660622
Molecular Formula: C6H7BrN2
Molecular Weight: 187.04
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 64


Other Product Names / Synonyms: 1,2-Diamino-4-bromobenzene

Formula: C₆H₇BrN₂
Melting Pt: 64 °C
MDL Number: MFCD02660622
CAS Number: 1575-37-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8fa446484cea2ea6b0dd4910d4ddc73ff29b4c4&vid=d8fa446484cea2ea6b0dd4910d4ddc73ff29b4c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR