7-Bromo-3-hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3180-1G B3180-5G
TCB3180-1GEA 47.4 CAD
TCB3180-1G TCB3180-5G
7-Bromo-3-hydroxy-2-naphthoic acid ≥98.0% (by GC, titration analysis)
7-Bromo-3-hydroxy-2-naphthoic acid

CAS Number: 1779-11-9
MDL Number: MFCD00137823
Molecular Formula: C11H7BrO3
Molecular Weight: 267.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 262


Other Product Names / Synonyms: 7-Bromo-3-hydroxy-2-naphthalenecarboxylic Acid

Formula: C₁₁H₇BrO₃
Melting Pt: 262 °C
MDL Number: MFCD00137823
CAS Number: 1779-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b34eb196f9f68494e9e95ca43a8da935ca4a6ae&vid=1b34eb196f9f68494e9e95ca43a8da935ca4a6ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR