O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine ≥98.0%
Supplier: TCI America
Synonyms:
N-Fmoc-O-tert-butyl-L-serine, N-a-Fmoc-O-t-butyl-L-serine, Fmoc-Ser(tBu)-OH, O-tert-Butyl-N-Fmoc-L-serine
CAS Number: 71989-33-8
MDL Number: MFCD00037127
Molecular Formula: C22H25NO5
Molecular Weight: 383.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 136
Specific rotation [a]20/D: 24 deg (C=1, EtOAc)
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: O-tert-Butyl-N-Fmoc-L-serine, Fmoc-Ser(tBu)-OH
Formula:
C₂₂H₂₅NO₅ MW: 383.44 g/mol Melting Pt: 136 °C |
MDL Number:
MFCD00037127 CAS Number: 71989-33-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...