1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B3158-1G
B3158-5G
TCB3158-1GEA
113.76
CAD
TCB3158-1G
TCB3158-5G
1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride ≥98.0% (by HPLC, titration analysis)
1,3-Bis(2,4,6-trimethylphenyl)imidazolinium Chloride
Formula:
C₂₁H₂₇ClN₂ Melting Pt: 286 °C |
MDL Number:
MFCD09039279 CAS Number: 173035-10-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...