1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride ≥96.0%
Supplier: TCI America
Synonyms:
SIPr.HCl, 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium chloride, 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride
CAS Number: 258278-25-0
MDL Number: MFCD07369796
Molecular Formula: C27H39ClN2
Molecular Weight: 427.07
Purity/Analysis Method: >96.0% (HPLC,N)
Form: Crystal
Melting point (°C): 293
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: SIPr.HCl
Formula:
C₂₇H₃₉ClN₂ MW: 427.07 g/mol Melting Pt: 293 °C |
MDL Number:
MFCD07369796 CAS Number: 258278-25-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...