(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98,0

Supplier: TCI America

Synonyms: N-Fmoc-L-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-L-aspartate, Fmoc-Asp(OtBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-tert-Butyl Ester

B3150-5G B3150-25G
TCB3150-5GEA 41.79 CAD
TCB3150-5G TCB3150-25G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98,0
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

CAS Number: 71989-14-5
MDL Number: MFCD00037131
Molecular Formula: C23H25NO6
Molecular Weight: 411.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: -25 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-tert-Butyl N-Fmoc-L-aspartate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-L-aspartic Acid 4-tert-Butyl Ester, Fmoc-Asp(OtBu)-OH

Formula: C₂₃H₂₅NO₆
MW: 411.5 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00037131
CAS Number: 71989-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb4f45a31985792b3eb48dd991fdc37abd332acd&vid=eb4f45a31985792b3eb48dd991fdc37abd332acd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR