2-Butylimidazole ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B3136-25G B3136-5G
TCB3136-25GEA 595.03 CAD
TCB3136-25G TCB3136-5G
2-Butylimidazole ≥97.0% (by GC, titration analysis)
2-Butylimidazole

CAS Number: 50790-93-7
MDL Number: MFCD00792664
Molecular Formula: C7H12N2
Molecular Weight: 124.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 294
Melting point (°C): 52

Formula: C₇H₁₂N₂
MW: 124.1800 g/mol
Boiling Pt: 294 °C
Melting Pt: 52 °C
MDL Number: MFCD00792664
CAS Number: 50790-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fe6aded64a2ccda4eda4e66b395186a529ffe4e&vid=1fe6aded64a2ccda4eda4e66b395186a529ffe4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR