4-(4-Bromophenyl)butyric acid ≥98.0%

Supplier: TCI America

Synonyms: 4-(4-Bromophenyl)butanoic acid

B3131-5G B3131-1G
TCB3131-5GEA 240.16 CAD
TCB3131-5G TCB3131-1G
4-(4-Bromophenyl)butyric acid ≥98.0%
4-(4-Bromophenyl)butyric acid

CAS Number: 35656-89-4
MDL Number: MFCD00463170
Molecular Formula: C10H11BrO2
Molecular Weight: 243.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 176
Melting point (°C): 72

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Boiling Pt: 176 °C
Melting Pt: 72 °C
MDL Number: MFCD00463170
CAS Number: 35656-89-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2daf4321aa99e9edca4a751ad52ca2d1f02d6d45&vid=2daf4321aa99e9edca4a751ad52ca2d1f02d6d45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR