6-Bromo-o-cresol ≥94.0%

Supplier: TCI America

Synonyms: 2-Bromo-6-methylphenol

B3126-5G B3126-25G
TCB3126-5GEA 281.24 CAD
TCB3126-5G TCB3126-25G
6-Bromo-o-cresol ≥94.0%
6-Bromo-o-cresol

CAS Number: 13319-71-6
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Melting point (°C): 16
Specific Gravity (20/20): 1.51


Other Product Names / Synonyms: 2-Bromo-6-methylphenol

Formula: C₇H₇BrO
MW: 187.0300 g/mol
Boiling Pt: 208 °C
Melting Pt: 16 °C
Density: 1.51 (20 °C)
MDL Number: MFCD09907876
CAS Number: 13319-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3d96205988738e7e722fde9e7a74c6a971a7c35&vid=c3d96205988738e7e722fde9e7a74c6a971a7c35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR