N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine ≥99.0% (by HPLC)
Supplier: TCI America
B3103-100MG
B3103-1G
TCB3103-100MGEA
148.84
CAD
TCB3103-100MG
TCB3103-1G
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine ≥99.0% (by HPLC)
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine
CAS Number: 64325-78-6
MDL Number: MFCD00010058
Molecular Formula: C38H35N5O6
Molecular Weight: 657.73
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 94
Specific rotation [a]20/D: -10 deg (C=1, MeOH)
Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: Bz-DMT-dA
Formula:
C₃₈H₃₅N₅O₆ Melting Pt: 94 °C |
MDL Number:
MFCD00010058 CAS Number: 64325-78-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...