N4-Benzoyl-2'-deoxycytidine ≥98.0%

Supplier: TCI America

Synonyms: N-(1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

B3102-100MG B3102-1G
TCB3102-100MGEA 30.02 CAD
TCB3102-100MG TCB3102-1G
N4-Benzoyl-2'-deoxycytidine ≥98.0%
N4-Benzoyl-2'-deoxycytidine

CAS Number: 4836-13-9
MDL Number: MFCD00010115
Molecular Formula: C16H17N3O5
Molecular Weight: 331.33
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 207
Specific rotation [a]20/D: 90 deg (C=0.25, MeOH)

Formula: C₁₆H₁₇N₃O₅
Melting Pt: 207 °C
MDL Number: MFCD00010115
CAS Number: 4836-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e177b95e1a06aff1dc55d0399eb53a06391be1d&vid=5e177b95e1a06aff1dc55d0399eb53a06391be1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR