N4-Benzoylcytidine 98,0

Supplier: TCI America

Synonyms: N-Benzoylcytidine

B3094-1G B3094-5G
TCB3094-1GEA 42.66 CAD
TCB3094-1G TCB3094-5G
N4-Benzoylcytidine 98,0
N4-Benzoylcytidine

CAS Number: 13089-48-0
MDL Number: MFCD00010572
Molecular Formula: C16H17N3O6
Molecular Weight: 347.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 240
Specific rotation [a]20/D: 46 deg (C=1, DMSO)

Formula: C₁₆H₁₇N₃O₆
MW: 347.33 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00010572
CAS Number: 13089-48-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR