4-Bromo-4',4''-dimethyltriphenylamine ≥95.0%

Supplier: TCI America

Synonyms: 4-Bromo-4,4-dimethyltriphenylamine, 4-Bromo-N,N-di-p-tolylaniline

B3089-1G B3089-5G
TCB3089-1GEA 100.8 CAD
TCB3089-1G TCB3089-5G
4-Bromo-4',4''-dimethyltriphenylamine ≥95.0%
4-Bromo-4',4''-dimethyltriphenylamine

CAS Number: 58047-42-0
Molecular Formula: C20H18BrN
Molecular Weight: 352.28
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 102


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 4-Bromo-N,N-di-p-tolylaniline

Formula: C₂₀H₁₈BrN
MW: 352.28 g/mol
Melting Pt: 102 °C
MDL Number: MFCD03093258
CAS Number: 58047-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d1a9211a5090290d22396d1cae1426ea5d77b21&vid=1d1a9211a5090290d22396d1cae1426ea5d77b21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR