N-Benzyl-2-naphthylamine ≥98.0%

Supplier: TCI America

Synonyms: N-Benzylnaphthalen-2-amine

B3088-25G B3088-5G
TCB3088-25GEA 1393.56 CAD
TCB3088-25G TCB3088-5G
N-Benzyl-2-naphthylamine ≥98.0%
N-Benzyl-2-naphthylamine

CAS Number: 13672-18-9
MDL Number: MFCD00144848
Molecular Formula: C17H15N
Molecular Weight: 233.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 68

Formula: C₁₇H₁₅N
MW: 233.31 g/mol
Boiling Pt: 190 °C
Melting Pt: 68 °C
MDL Number: MFCD00144848
CAS Number: 13672-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a5225b1b7d7f7dd7b5e39d06a6d5772882579e9&vid=7a5225b1b7d7f7dd7b5e39d06a6d5772882579e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR