N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0%

Supplier: TCI America

Synonyms: Bz-DMT-dC, N-(1-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

B3087-5G B3087-1G
TCB3087-5GEA 690.93 CAD
TCB3087-5G TCB3087-1G
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0%
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine

CAS Number: 67219-55-0
MDL Number: MFCD00010114
Molecular Formula: C37H35N3O7
Molecular Weight: 633.70
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 119
Specific rotation [a]20/D: 58 deg (C=1, MeOH)


Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: Bz-DMT-dC

Formula: C₃₇H₃₅N₃O₇
MW: 633.70 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00010114
CAS Number: 67219-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6cc3f40bc4ac95dc26ae3852aa10f91e87e33dc&vid=e6cc3f40bc4ac95dc26ae3852aa10f91e87e33dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR