Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98.0%

Supplier: TCI America

Synonyms: Nɑ-Boc-D-lysine, Boc-D-Lys-OH

B3083-5G B3083-1G
TCB3083-5GEA 188.49 CAD
TCB3083-5G TCB3083-1G
Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98.0%
Nalpha-(tert-Butoxycarbonyl)-D-lysine

CAS Number: 106719-44-2
MDL Number: MFCD00076956
Molecular Formula: C11H22N2O4
Molecular Weight: 246.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 198
Specific rotation [a]20/D: -22 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-D-lysine, Boc-D-Lys-OH

Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00076956
CAS Number: 106719-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d95ba06b7b35524649761c63b30b68083503d9c&vid=6d95ba06b7b35524649761c63b30b68083503d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR