2-Bromo-1,3-difluoro-5-iodobenzene ≥98.0%

Supplier: TCI America

B3073-5G B3073-25G
TCB3073-5GEA 106.13 CAD
TCB3073-5G TCB3073-25G
2-Bromo-1,3-difluoro-5-iodobenzene ≥98.0%
2-Bromo-1,3-difluoro-5-iodobenzene

CAS Number: 155906-10-8
MDL Number: MFCD06797752
Molecular Formula: C6H2BrF2I
Molecular Weight: 318.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 66

Formula: C₆H₂BrF₂I
Melting Pt: 66 °C
MDL Number: MFCD06797752
CAS Number: 155906-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a5d5d3efc585b1893ec2a68b16f08fbe5811d2c&vid=8a5d5d3efc585b1893ec2a68b16f08fbe5811d2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR