1-Bromo-3-fluoro-4-nitrobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-2-fluoronitrobenzene

B3062-25G B3062-5G
TCB3062-25GEA 279.66 CAD
TCB3062-25G TCB3062-5G
1-Bromo-3-fluoro-4-nitrobenzene ≥98.0%
1-Bromo-3-fluoro-4-nitrobenzene

CAS Number: 321-23-3
MDL Number: MFCD01930221
Molecular Formula: C6H3BrFNO2
Molecular Weight: 220.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86

Formula: C₆H₃BrFNO₂
Melting Pt: 86 °C
MDL Number: MFCD01930221
CAS Number: 321-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf57a070d48f6791b4e21fdb8a1cefa32eb1897d&vid=bf57a070d48f6791b4e21fdb8a1cefa32eb1897d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR