4-Bromo-2-chlorobenzotrifluoride ≥98.0%

Supplier: TCI America

B3057-25G B3057-5G
TCB3057-25GEA 269.39 CAD
TCB3057-25G TCB3057-5G
4-Bromo-2-chlorobenzotrifluoride ≥98.0%
4-Bromo-2-chlorobenzotrifluoride

CAS Number: 467435-07-0
MDL Number: MFCD08459292
Molecular Formula: C7H3BrClF3
Molecular Weight: 259.45
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.76


Other Product Names / Synonyms: 4-Bromo-2-chloro-1-(trifluoromethyl)benzene, 4-Bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Density: 1.76 (20 °C)
MDL Number: MFCD08459292
CAS Number: 467435-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2dcfd98b2046ab11b7df40c7617349dbc32adc2&vid=f2dcfd98b2046ab11b7df40c7617349dbc32adc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR