N-Benzyl-N-nitroso-p-toluenesulfonamide ≥98.0% (by HPLC)

Supplier: TCI America

B3051-25G B3051-5G
TCB3051-25GEA 986.87 CAD
TCB3051-25G TCB3051-5G
N-Benzyl-N-nitroso-p-toluenesulfonamide ≥98.0% (by HPLC)
N-Benzyl-N-nitroso-p-toluenesulfonamide

CAS Number: 33528-13-1
MDL Number: MFCD00216617
Molecular Formula: C14H14N2O3S
Molecular Weight: 290.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 85
Storage Temperature: 0-10°C

Formula: C₁₄H₁₄N₂O₃S
Melting Pt: 85 °C
MDL Number: MFCD00216617
CAS Number: 33528-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c692c739ccb97807021b2f7376995ca3be529cc7&vid=c692c739ccb97807021b2f7376995ca3be529cc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR