5-Bromo-o-toluic acid ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-2-methylbenzoic acid

B3050-25G B3050-5G
TCB3050-25GEA 80.58 CAD
TCB3050-25G TCB3050-5G
5-Bromo-o-toluic acid ≥98.0%
5-Bromo-o-toluic acid

CAS Number: 79669-49-1
MDL Number: MFCD00267350
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 171


Other Product Names / Synonyms: 5-Bromo-o-toluic Acid

Formula: C₈H₇BrO₂
Melting Pt: 171 °C
MDL Number: MFCD00267350
CAS Number: 79669-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b044f9a68ac2703a1ec63af822162be143f4b10d&vid=b044f9a68ac2703a1ec63af822162be143f4b10d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR