trans-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3037-5G
B3037-1G
TCB3037-5GEA
507.18
CAD
TCB3037-5G
TCB3037-1G
trans-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-tert-Butylcyclohexanecarboxylic acid
CAS Number: 943-29-3
MDL Number: MFCD00466237
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 176
Formula:
C₁₁H₂₀O₂ Melting Pt: 176 °C |
MDL Number:
MFCD00466237 CAS Number: 943-29-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...